2-Bromononane ≥97%
Supplier: Thermo Fisher Scientific
L07387.03
L07387.06
L07387.03EA
37
EUR
L07387.03
L07387.06
2-Bromononane ≥97%
2-Bromononane
Purity: 97%
CAS No.: 2216-35-5
Molecular Formula: C9H19Br
Formula Weight: 207.16
Refractive Index: 1.4530
Boiling Point: 208-209°
MDL No.: MFCD00017865
Formula:
H₃CBrCH(CH₂)₆CH₃ MW: 207.15 g/mol Boiling Pt: 208…209 °C Density: 1.081 Storage Temperature: Ambient |
MDL Number:
MFCD00017865 CAS Number: 2216-35-5 EINECS: 218-688-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...