Bromoethane 98%, pure
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Bromic ether, Hydrobromic ether
154210010
154215000
154210025
ACRO154210010EA
95.8
EUR
ACRO154210010
ACRO154215000
ACRO154210025
Bromoethane 98%, pure
Bromoethane
Formula:
BrH₂CCH₃ MW: 108.97 g/mol Boiling Pt: 38.4 Melting Pt: –119 °C Density: 1.46 g/cm³ Flash Pt: –23 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000232 CAS Number: 74-96-4 EINECS: 200-825-8 UN: 1891 ADR: 6.1 II Merck Index: 13,03807 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4230 to 1.4250 (20°C, 589 nm) |
Water | ≤0.03 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...