Bromoethane for synthesis, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.00871.0250 8.00871.1000
8.00871.0250EA 33.3 EUR
8.00871.0250 8.00871.1000
Bromoethane for synthesis, Sigma-Aldrich®
Bromoethane
Formula: BrH₂CCH₃
Boiling Pt: 38,4 °C (1013 hPa)
Melting Pt: –118 °C
Density: 1,46 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00000232
CAS Number: 74-96-4
EINECS: 200-825-8
UN: 1891
ADR: 6.1,II
Merck Index: 13,03807

Order Now

Specification Test Results

Solubility 9 g/l (20 °C)
Melting Point -118 °C
Molar Mass 108.96 g/mol
Boiling Point 37 - 39 °C (1013 hPa)
Vapor Pressure 510 hPa (20 °C)
Flash Point -20 °C
Refractive Index 1.4236 (20 °C, 589 nm)
Explosion Limit 6.7 - 11.3 %(V)
Density 1.46 g/cm3 (20 °C)
Ignition Point 510 °C
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR