Dibromochloromethane 98%, rods

Supplier: Thermo Fisher Scientific
Warning

A16938.06 A16938.14
A16938.06EA 39.6 EUR
A16938.06 A16938.14
Dibromochloromethane 98%, rods
Dibromochloromethane

Purity: 98% CAS No.: 124-48-1 Molecular Formula: ClCHBr2 Formula Weight: 208.28 Storage Temperature: 2°C to 8°C Refractive Index: 1.5475 Melting Point: -22° Boiling Point: 115-118° Beilstein Reference: 1731046 Merck Reference: 14,2135 MDL No.: MFCD00000820

Formula: Br₂ClCH
Boiling Pt: 115…118 °C
Melting Pt: –22 °C
Density: 2,420
Storage Temperature: Refrigerator
MDL Number: MFCD00000820
CAS Number: 124-48-1
EINECS: 204-704-0
Merck Index: 14,2135

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR