Heptanal ≥97%

Supplier: Thermo Fisher Scientific
Warning

B23830.AE B23830.AK B23830.AP
B23830.AEEA 28.4 EUR
B23830.AE B23830.AK B23830.AP
Heptanal ≥97%
Heptanal

Purity: 97% CAS No.: 111-71-7 Molecular Formula: CH3(CH2)5CHO Formula Weight: 114.19 Flash Point: 35°(95°F) Physical Form: Liquid Density: 0.850 Refractive Index: 1.4125 Sensitivity: Hygroscopic Odour: Unpleasant Melting Point: -43° Boiling Point: 152-154° Beilstein Reference: 1560236 Merck Reference: 14,4658 UN No.: 3056 MDL No.: MFCD00007028

Formula: C₇H₁₄O
Boiling Pt: 152…154 °C
Melting Pt: –43 °C
Density: 0,850
Flash Pt: 35 °C (95 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007028
CAS Number: 111-71-7
EINECS: 203-898-4
UN: 3056
ADR: 3,III
Merck Index: 14,4658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR