Undecanal ≥97%
Supplier: Thermo Scientific
Warning
|
Synonyms:
Hendecanal, Undecylaldehyde
A16101.14
A16101.22
A16101.14EA
43.8
EUR
A16101.14
A16101.22
Undecanal ≥97%
Undecanal
Purity: 97%
CAS No.: 112-44-7
Molecular Formula: C11H22O
Formula Weight: 170.30
Flash Point: 95°(203°F)
Refractive Index: 1.4340
Melting Point: -2°
Boiling Point: 110-112°/12mm
Beilstein Reference: 1753213
MDL No.: MFCD00007016
Formula:
C₁₁H₂₂O MW: 170,3 g/mol Boiling Pt: 110…112 °C (12 mmHg) Melting Pt: –2 °C Density: 0,827 Flash Pt: 95 °C (203 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007016 CAS Number: 112-44-7 EINECS: 203-972-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...