L(-)-Camphorsulphonic acid 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(1R)-(-)-10-Camphorsulphonic acid, ((1R,4S)-7,7-Dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl)methanesulphonic acid, (-)-10-Camphorsulphonic acid, (-)-CSA
263640250
263640050
263641000
263645000
ACRO263640250EA
50.4
EUR
ACRO263640250
ACRO263640050
ACRO263641000
ACRO263645000
L(-)-Camphorsulphonic acid 98%
L(-)-Camphorsulphonic acid
Formula:
C₁₀H₁₆O₄S MW: 232,3 g/mol Boiling Pt: 375 °C (1013 hPa) Melting Pt: 198 °C (dec.) Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064158 CAS Number: 35963-20-3 EINECS: 252-817-9 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Specific optical rotation | -21° to -23° (20°C, 589 nm) (c=20, H2O) |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...