2-(2-Bromoethoxy)anisole ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 1-(2-Bromoethoxy)-2-methoxybenzene

H52382.MD H52382.06
H52382.MDEA 0 EUR
H52382.MD H52382.06
2-(2-Bromoethoxy)anisole ≥97%
2-(2-Bromoethoxy)anisole

Purity: 97+% CAS No.: 4463-59-6 Molecular Formula: C9H11BrO2 Formula Weight: 231.09 MDL No.: MFCD00017892

Formula: C₉H₁₁BrO₂
MW: 231,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017892
CAS Number: 4463-59-6
EINECS: 402-010-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR