3,4-Diaminobenzophenone ≥97%
Supplier: Thermo Fisher Scientific
A13863.06
A13863.14
A13863.06EA
0
EUR
A13863.06
A13863.14
3,4-Diaminobenzophenone ≥97%
3,4-Diaminobenzophenone
Purity: 97%
CAS No.: 39070-63-8
Molecular Formula: C13H12N2O
Formula Weight: 212.25
Melting Point: 115-119°
Beilstein Reference: 2212487
Formula:
C₁₃H₁₂N₂O Boiling Pt: 390 °C (1013 hPa) Melting Pt: 116…118 °C |
MDL Number:
MFCD00007727 CAS Number: 39070-63-8 EINECS: 254-273-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...