3,5-Difluoro-4-iodophenol ≥98%
Supplier: Thermo Scientific
H32537.03
H32537.06
H32537.03EA
0
EUR
H32537.03
H32537.06
3,5-Difluoro-4-iodophenol ≥98%
3,5-Difluoro-4-iodophenol
Purity: 98%
CAS No.: 1000574-74-2
Molecular Formula: C6H3F2IO
Formula Weight: 255.99
Physical Form: Crystalline powder
Sensitivity: Light Sensitive
Appearance: White to pale yellow
Melting Point: 103-112°
MDL No.: MFCD09955025
Formula:
C₆H₃F₂IO MW: 255,99 g/mol Melting Pt: 103…112 °C |
MDL Number:
MFCD09955025 CAS Number: 1000574-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...