2-Amino-5-nitrobenzophenone 98%

Supplier: Thermo Scientific
Warning

Synonyms: (2-Amino-5-nitrophenyl)(phenyl)methanone

B23773.06 B23773.22
B23773.06EA 0 EUR
B23773.06 B23773.22
2-Amino-5-nitrobenzophenone 98%
2-Amino-5-nitrobenzophenone

Purity: 98% CAS No.: 1775-95-7 Molecular Formula: C13H10N2O3 Formula Weight: 242.23 Melting Point: 166-168° MDL No.: MFCD00007364

Formula: C₁₃H₁₀N₂O₃
MW: 242,23 g/mol
Melting Pt: 166…168 °C
Density: 1,474 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007364
CAS Number: 1775-95-7
EINECS: 217-207-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR