1-Bromo-2-pentyne ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
H53433.06
H53433.14
H53433.06EA
107
EUR
H53433.06
H53433.14
1-Bromo-2-pentyne ≥97%
1-Bromo-2-pentyne
Purity: 97%
CAS No.: 16400-32-1
Molecular Formula: C5H7Br
Formula Weight: 147.01
Flash Point: 41°(106°F)
Refractive Index: 1.498
Boiling Point: 93-94°/113mm
UN No.: 1993
MDL No.: MFCD00236363
Formula:
C₅H₇Br MW: 147.01 g/mol Boiling Pt: 93…94 °C (113 mmHg) Density: 1.438 Flash Pt: 41 °C (106 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00236363 CAS Number: 16400-32-1 EINECS: 240-451-2 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...