1-Chloro-2,6-dibromo-4-fluorobenzene ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 1,3-Dibromo-2-chloro-5-fluorobenzene, 2-Chloro-1,3-dibromo-5-fluorobenzene

B25376.14
B25376.14EA 76.4 EUR
B25376.14
1-Chloro-2,6-dibromo-4-fluorobenzene ≥98%
1-Chloro-2,6-dibromo-4-fluorobenzene

Purity: 98% CAS No.: 179897-90-6 Molecular Formula: C6H2Br2ClF Formula Weight: 288.35 Melting Point: 84-87° MDL No.: MFCD00061122

Formula: C₆H₂Br₂ClF
MW: 288,34 g/mol
Melting Pt: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00061122
CAS Number: 179897-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR