2-Fluoro-5-(trifluoromethoxy)phenol ≥97%

Supplier: Thermo Fisher Scientific
Danger

H26535.03 H26535.06
H26535.03EA 0 EUR
H26535.03 H26535.06
2-Fluoro-5-(trifluoromethoxy)phenol ≥97%
2-Fluoro-5-(trifluoromethoxy)phenol

Purity: 97% CAS No.: 886498-03-9 Molecular Formula: C7H4F4O2 Formula Weight: 196.10 MDL No.: MFCD04115893

Formula: C₇H₄F₄O₂
Storage Temperature: Ambient
MDL Number: MFCD04115893
CAS Number: 886498-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR