5-Fluoro-2-(trifluoromethyl)phenol ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Fluoro-2-hydroxybenzotrifluoride

H32445.03 H32445.06
H32445.03EA 0 EUR
H32445.03 H32445.06
5-Fluoro-2-(trifluoromethyl)phenol ≥97%
5-Fluoro-2-(trifluoromethyl)phenol

Purity: 97% CAS No.: 243459-91-8 Molecular Formula: C7H4F4O Formula Weight: 180.10 Melting Point: 48-52° UN No.: 3261 MDL No.: MFCD00236302

Formula: C₇H₄F₄O
MW: 180.1 g/mol
Melting Pt: 48…52 °C
Storage Temperature: Ambient
MDL Number: MFCD00236302
CAS Number: 243459-91-8
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR