3',4'-Dihydroxyflavone ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(3,4-Dihydroxyphenyl)-4-benzopyrone, 2-(3,4-Dihydroxyphenyl)chromen-4-one
L14161.MC
L14161.ME
L14161.MCEA
74.7
EUR
L14161.MC
L14161.ME
3',4'-Dihydroxyflavone ≥97%
3',4'-Dihydroxyflavone
Purity: 97%
CAS No.: 4143-64-0
Molecular Formula: C15H10O4
Formula Weight: 254.24
Melting Point: 249-252°
Beilstein Reference: 212540
Formula:
C₁₅H₁₀O₄ MW: 254,24 g/mol Melting Pt: 249…252 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017600 CAS Number: 4143-64-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...