2-(Chloromethyl)benzimidazole 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(Chloromethyl)-1H-benzimidazole
B25548.06
B25548.14
B25548.06EA
49.6
EUR
B25548.06
B25548.14
2-(Chloromethyl)benzimidazole 95%
2-(Chloromethyl)benzimidazole
Purity: 96%
CAS No.: 4857-04-9
Molecular Formula: C8H7ClN2
Formula Weight: 166.61
Melting Point: 141-143° dec.
MDL No.: MFCD00005602
Formula:
C₈H₇ClN₂ MW: 166,61 g/mol Melting Pt: 141…143 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005602 CAS Number: 4857-04-9 EINECS: 225-452-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...