1,1,1-Tris(hydroxymethyl)ethane 97%

Supplier: Thermo Fisher Scientific
Warning

140510250 140515000 140510025
ACRO140510250EA 25.8 EUR
ACRO140510250 ACRO140515000 ACRO140510025
1,1,1-Tris(hydroxymethyl)ethane 97%
1,1,1-Tris(hydroxymethyl)ethane
Formula: C₅H₁₂O₃
Boiling Pt: 287 °C (1013 hPa)
Melting Pt: 200…203 °C
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00004687
CAS Number: 77-85-0
EINECS: 201-063-9

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR