2,2,4,4-Tetramethyl-3-pentanone ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Di-tert-butyl ketone, Hexamethylacetone
H53464.06
H53464.14
H53464.06EA
54.3
EUR
H53464.06
H53464.14
2,2,4,4-Tetramethyl-3-pentanone ≥98%
2,2,4,4-Tetramethyl-3-pentanone
Purity: 98%
CAS No.: 815-24-7
Molecular Formula: C9H18O
Formula Weight: 142.24
Flash Point: 34°(93°F)
Refractive Index: 1.419
Boiling Point: 152-153°
Beilstein Reference: 1701122
UN No.: 1224
MDL No.: MFCD00008845
Formula:
C₉H₁₈O MW: 142.24 g/mol Boiling Pt: 152…153 °C Melting Pt: –25.2 °C Density: 0.824 Flash Pt: 34 °C (93 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008845 CAS Number: 815-24-7 EINECS: 212-419-8 UN: 1224 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...