2,5-Dibromo-4-picoline ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2,5-Dibromo-4-methylpyridine
H30457.03
H30457.06
H30457.03EA
0
EUR
H30457.03
H30457.06
2,5-Dibromo-4-picoline ≥97%
2,5-Dibromo-4-picoline
Purity: 97%
CAS No.: 3430-26-0
Molecular Formula: C6H5Br2N
Formula Weight: 250.92
Melting Point: 36-40°
MDL No.: MFCD00234955
Formula:
C₆H₅Br₂N MW: 250.92 g/mol Melting Pt: 36…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234955 CAS Number: 3430-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...