3,6-Difluoro-2-methoxybenzaldehyde ≥97%

Warning

H32549.MD H32549.03
H32549.MDEA 88.7 EUR
H32549.MD H32549.03
3,6-Difluoro-2-methoxybenzaldehyde ≥97%
3,6-Difluoro-2-methoxybenzaldehyde

Purity: 97+% CAS No.: 887267-04-1 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Melting Point: 38-45° MDL No.: MFCD08275289

Formula: C₈H₆F₂O₂
MW: 172,13 g/mol
Melting Pt: 38…45 °C
Storage Temperature: Ambient
MDL Number: MFCD08275289
CAS Number: 887267-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR