2-Bromo-4-(trifluoromethyl)benzenesulphonyl chloride ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Bromo-4-(trifluoromethyl)benzene-1-sulphonyl chloride

L19712.03 L19712.06
L19712.03EA 46.2 EUR
L19712.03 L19712.06
2-Bromo-4-(trifluoromethyl)benzenesulphonyl chloride ≥97%
2-Bromo-4-(trifluoromethyl)benzenesulphonyl chloride

Purity: 97% CAS No.: 54403-98-4 Molecular Formula: C7H3BrClF3O2S Formula Weight: 323.52 Refractive Index: 1.5270 Boiling Point: 235-236° UN No.: 3265 MDL No.: MFCD03094394

Formula: C₇H₃BrClF₃O₂S
MW: 323,52 g/mol
Boiling Pt: 235…236 °C
Density: 1,865
Storage Temperature: Ambient
MDL Number: MFCD03094394
CAS Number: 54403-98-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR