6-Bromo-m-toluic acid ≥98%
Supplier: Thermo Scientific
|
Warning
|
H32927.03
H32927.06
H32927.03EA
24.5
EUR
H32927.03
H32927.06
6-Bromo-m-toluic acid ≥98%
6-Bromo-m-toluic acid
Purity: 98%
CAS No.: 6967-82-4
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
MDL No.: MFCD00079722
Formula:
C₈H₇BrO₂ MW: 215,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00079722 CAS Number: 6967-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...