2-Bromobenzimidazole ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Bromo-1H-benzimidazole

H26183.03 H26183.06
H26183.03EA 82.6 EUR
H26183.03 H26183.06
2-Bromobenzimidazole ≥99%
2-Bromobenzimidazole

Purity: 99% CAS No.: 54624-57-6 Molecular Formula: C7H5BrN2 Formula Weight: 197.03 Melting Point: 195-201° MDL No.: MFCD00160009

Formula: C₇H₅BrN₂
Melting Pt: 195…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00160009
CAS Number: 54624-57-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR