1,1,1-Tris(hydroxymethyl)propane 98%

Supplier: Thermo Fisher Scientific

164650010 164650025 164655000 164650100
ACRO164650010EA 67 EUR
ACRO164650010 ACRO164650025 ACRO164655000 ACRO164650100
1,1,1-Tris(hydroxymethyl)propane 98%
1,1,1-Tris(hydroxymethyl)propane
Formula: C₆H₁₄O₃
Boiling Pt: 160 °C (,7hPa)
Melting Pt: 56…60 °C
Density: 1,15 g/cm³ (20 °C)
Flash Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD00004694
CAS Number: 77-99-6
EINECS: 201-074-9

Order Now

Specification Test Results

Appearance White Flakes
Infrared spectrum Conforms
Melting point ≥57 °C
Water ≤0.1 %
Ash ≤0.0015 %
Acidity ≤0.002 % (as Formic acid)
Hydroxyl value ≥1235 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR