2-Fluoro-5-methoxybenzoic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
H52365.MD
H52365.03
H52365.06
H52365.MDEA
84.4
EUR
H52365.MD
H52365.03
H52365.06
2-Fluoro-5-methoxybenzoic acid ≥97%
2-Fluoro-5-methoxybenzoic acid
Purity: 97+%
CAS No.: 367-83-9
Molecular Formula: C8H7FO3
Formula Weight: 170.14
MDL No.: MFCD00272566
Formula:
C₈H₇FO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00272566 CAS Number: 367-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...