2-Hydroxy-5-nitro-4-picoline ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Hydroxy-4-methyl-5-nitropyridine, 4-Methyl-5-nitro-2-pyridone

L19867.03
L19867.03EA 44 EUR
L19867.03
2-Hydroxy-5-nitro-4-picoline ≥97%
2-Hydroxy-5-nitro-4-picoline

Purity: 97% CAS No.: 21901-41-7 Molecular Formula: C6H6N2O3 Formula Weight: 154.13 Odour: Odourless Melting Point: 188-190° Beilstein Reference: 136900 MDL No.: MFCD00010690

Formula: C₆H₆N₂O₃
MW: 154,13 g/mol
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00010690
CAS Number: 21901-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR