2-Mercapto-5-nitrobenzimidazole ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Nitrobenzimidazole-2-thiol, 2-Mercapto-5-nitro-1H-benzimidazole
B22358.06
B22358.14
B22358.06EA
37.3
EUR
B22358.06
B22358.14
2-Mercapto-5-nitrobenzimidazole ≥97%
2-Mercapto-5-nitrobenzimidazole
Purity: 97%
CAS No.: 6325-91-3
Molecular Formula: C7H5N3O2S
Formula Weight: 195.20
Melting Point: ca 273° dec.
Beilstein Reference: 172127
Formula:
C₇H₅N₃O₂S MW: 195,2 g/mol Melting Pt: ∼ 273 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00804727 CAS Number: 6325-91-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...