2-Mercaptoimidazole ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Imidazole-2-thiol, 2-Mercapto-1H-imidazole

L01346.03 L01346.06 L01346.14
L01346.03EA 55 EUR
L01346.03 L01346.06 L01346.14
2-Mercaptoimidazole ≥98%
2-Mercaptoimidazole

Purity: 98+% CAS No.: 872-35-5 Molecular Formula: C3H4N2S Formula Weight: 100.14 Melting Point: 222-227° Beilstein Reference: 105775

Formula: C₃H₄N₂S
MW: 100,14 g/mol
Melting Pt: 222…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00005188
CAS Number: 872-35-5
EINECS: 212-823-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR