2-Methoxythiobenzamide ≥97%
Supplier: Thermo Fisher Scientific
|
Danger
|
H32737.03
H32737.06
H32737.03EA
81.2
EUR
H32737.03
H32737.06
2-Methoxythiobenzamide ≥97%
2-Methoxythiobenzamide
Purity: 97%
CAS No.: 42590-97-6
Molecular Formula: C8H9NOS
Formula Weight: 167.23
Melting Point: 109-113°
MDL No.: MFCD04973324
Formula:
C₈H₉NOS Melting Pt: 109…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973324 CAS Number: 42590-97-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...