Galangin ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3,5,7-Trihydroxy-2-phenyl-4H-benzo[b]pyran-4-one, 3,5,7-Trihydroxy-2-phenyl-4-benzopyrone, Norizalpinin, 3,5,7-Trihydroxyflavone, 3,5,7-Trihydroxy-2-phenylchromen-4-one, 3,5,7-Trihydroxy-2-phenyl-4H-chromen-4-one, 3,5,7-Trihydroxy-2-phenyl-4H-1-benzopyran-4-one

L14223.MB L14223.MC
L14223.MBEA 139 EUR
L14223.MB L14223.MC
Galangin ≥97%
Galangin

Purity: 97% CAS No.: 548-83-4 Molecular Formula: C15H10O5 Formula Weight: 270.24 Melting Point: 214-216° Beilstein Reference: 272179 Merck Reference: 14,4339 MDL No.: MFCD00006833

Formula: C₁₅H₁₀O₅
MW: 270,24 g/mol
Melting Pt: 214…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00006833
CAS Number: 548-83-4
EINECS: 208-960-4
Merck Index: 14,4339

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR