α-Naphthoflavone 97%
Supplier: Thermo Scientific
Synonyms:
2-Phenyl-4H-benzo[h]chromen-4-one, 2-Phenylbenzo[h]chromen-4-one, 7,8-Benzoflavone
128160010
128160050
ACRO128160010EA
34.5
EUR
ACRO128160010
ACRO128160050
α-Naphthoflavone 97%
α-Naphthoflavone
Formula:
C₁₉H₁₂O₂ MW: 272,3 g/mol Boiling Pt: 431 °C (1013 hPa) Melting Pt: 155…158 °C |
MDL Number:
MFCD00004985 CAS Number: 604-59-1 EINECS: 210-071-1 |
Specification Test Results
Appearance | Off-white to light yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 155 to 158 °C |
HPLC | ≥99.50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...