Flavone ≥99%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Phenylchromen-4-one

A13627.03 A13627.06 A13627.14
A13627.03EA 30.6 EUR
A13627.03 A13627.06 A13627.14
Flavone ≥99%
Flavone

Purity: 99% CAS No.: 525-82-6 Molecular Formula: C15H10O2 Formula Weight: 222.24 Odour: Odourless Melting Point: 95-98° Beilstein Reference: 157598 Merck Reference: 14,4092 MDL No.: MFCD00006825

Formula: C₁₅H₁₀O₂
MW: 222,24 g/mol
Boiling Pt: 180…185 °C (1 mmHg)
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00006825
CAS Number: 525-82-6
EINECS: 208-383-8
Merck Index: 14,4092

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR