3,4,5-Trihydroxybenzamide ≥98%
Supplier: Thermo Scientific
|
Warning
|
B20620.06
B20620.14
B20620.06EA
33.7
EUR
B20620.06
B20620.14
3,4,5-Trihydroxybenzamide ≥98%
3,4,5-Trihydroxybenzamide
Purity: 98%
CAS No.: 618-73-5
Molecular Formula: C7H7NO4
Formula Weight: 169.14
Melting Point: ca 240° dec.
Beilstein Reference: 2644549
Formula:
C₇H₇NO₄ Boiling Pt: 403 °C (1013 hPa) Melting Pt: ∼ 240 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00014800 CAS Number: 618-73-5 EINECS: 210-561-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...