3,5-Dinitrobenzamide ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Nitromide

B21865.14 B21865.22
B21865.14EA 77.2 EUR
B21865.14 B21865.22
3,5-Dinitrobenzamide ≥98%
3,5-Dinitrobenzamide

Purity: 98% CAS No.: 121-81-3 Molecular Formula: C7H5N3O5 Formula Weight: 211.13 Melting Point: 181-183° Merck Reference: 14,6612 MDL No.: MFCD00007985

Formula: C₇H₅N₃O₅
Melting Pt: 181…183 °C
Storage Temperature: Ambient
MDL Number: MFCD00007985
CAS Number: 121-81-3
EINECS: 204-499-8
Merck Index: 14,6612

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR