3-Chloro-2-fluorophenylboronic acid ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(3-Chloro-2-fluorophenyl)metaboric acid, Dihydroxy(3-chloro-2-fluorophenyl)borane, 3-Chloro-2-fluorophenylboronic Acid, 3-Chloro-2-fluorobenzeneboronic acid
H52665.03
H52665.06
H52665.03EA
37.8
EUR
H52665.03
H52665.06
3-Chloro-2-fluorophenylboronic acid ≥97%
3-Chloro-2-fluorophenylboronic acid
Purity: 97%
CAS No.: 352535-82-1
Molecular Formula: C6H5BClFO2
Formula Weight: 174.37
Melting Point: 240-242°
MDL No.: MFCD05664224
Formula:
C₆H₅BClFO₂ MW: 174,37 g/mol Melting Pt: 240…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664224 CAS Number: 352535-82-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...