3-Chloro-4-methylbenzenesulphonyl chloride ≥99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-Chlor-4-methylbenzenesulphonyl chloride, 2-Chloro-p-toluenesulphochloride, 3-Chloro-4-methylbenzene-1-sulphonyl chloride
L19886.03
L19886.06
L19886.14
L19886.03EA
51.7
EUR
L19886.03
L19886.06
L19886.14
3-Chloro-4-methylbenzenesulphonyl chloride ≥99%
3-Chloro-4-methylbenzenesulphonyl chloride
Purity: 99%
CAS No.: 42413-03-6
Molecular Formula: C7H6Cl2O2S
Formula Weight: 225.09
Flash Point:
Formula:
C₇H₆Cl₂O₂S MW: 225,09 g/mol Boiling Pt: 155…156 °C (19 mmHg) Melting Pt: 34…38 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00203987 CAS Number: 42413-03-6 EINECS: 412-890-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...