5-Chloro-2-fluorophenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Chloro-2-fluorobenzeneboronic acid, (5-Chloro-2-fluorophenyl)metaboric acid, (5-Chloro-2-fluorophenyl)boronic acid, Dihydroxy(5-chloro-2-fluorophenyl)borane

H53108.03 H53108.06
H53108.03EA 36.3 EUR
H53108.03 H53108.06
5-Chloro-2-fluorophenylboronic acid ≥97%
5-Chloro-2-fluorophenylboronic acid

Purity: 97% CAS No.: 352535-83-2 Molecular Formula: C6H5BClFO2 Formula Weight: 174.37 Melting Point: 182-188° MDL No.: MFCD05664225

Formula: C₆H₅BClFO₂
MW: 174,37 g/mol
Melting Pt: 182…188 °C
Storage Temperature: Ambient
MDL Number: MFCD05664225
CAS Number: 352535-83-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR