5-Fluoro-m-anisaldehyde ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Fluoro-5-methoxybenzaldehyde, 5-Fluoro-3-methoxybenzaldehyde, 3-Fluoro-5-anisaldehyde

H32415.03
H32415.03EA 187 EUR
H32415.03
5-Fluoro-m-anisaldehyde ≥98%
5-Fluoro-m-anisaldehyde

Purity: 98% CAS No.: 699016-24-5 Molecular Formula: C8H7FO2 Formula Weight: 154.14 Melting Point: 24-29° MDL No.: MFCD08741376

Formula: C₈H₇FO₂
MW: 154,14 g/mol
Melting Pt: 24…29 °C
Storage Temperature: Ambient
MDL Number: MFCD08741376
CAS Number: 699016-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR