3-Heptyne ≥97%

Supplier: Thermo Scientific
Danger

H53504.06 H53504.14
H53504.06EA 48 EUR
H53504.06 H53504.14
3-Heptyne ≥97%
3-Heptyne
Formula: C₇H₁₂
MW: 96,17 g/mol
Boiling Pt: 105…106 °C
Melting Pt: –130,5 °C
Density: 0,741
Flash Pt: –16 °C (3 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039960
CAS Number: 2586-89-2
EINECS: 219-967-7
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR