4-Isopropylbenzenesulphonyl chloride 96%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
4-(Methylethyl)benzolsulphonsäurechlorid, 4-Isopropylbenzene-1-sulphonyl chloride
L10428.06
L10428.14
L10428.06EA
51.9
EUR
L10428.06
L10428.14
4-Isopropylbenzenesulphonyl chloride 96%
4-Isopropylbenzenesulphonyl chloride
Purity: 96%
CAS No.: 54997-90-9
Molecular Formula: C9H11ClO2S
Formula Weight: 218.70
Flash Point:
Formula:
C₉H₁₁ClO₂S Boiling Pt: 142 °C (5 mmHg) Density: 1,220 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041510 CAS Number: 54997-90-9 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...