Luteolin

Supplier: Cayman Chemical
Warning

Synonyms: 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4-benzopyrone, Natural Yellow 2, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-4H-chromen-4-one, 3',4',5,7-Tetrahydroxyflavone

10004161-10 10004161-100 10004161-50 10004161-500
CAYM10004161-10EA 41.5 EUR
CAYM10004161-10 CAYM10004161-100 CAYM10004161-50 CAYM10004161-500
Luteolin
Luteolin
Formula: C₁₅H₁₀O₆
MW: 286.24 g/mol
MDL Number: MFCD00017309
CAS Number: 491-70-3
EINECS: 207-741-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR