Genistein

Supplier: Molekula

Synonyms: 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4-benzopyrone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-chromen-4-one, 4',6'-Dihydroxy-3-(4-hydroxyphenyl)benzo(e)oxacyclohex-2-en-4-on, 5,7-Dihydroxy-3-(4-hydroxyphenyl)chromen-4-one, 4',5,7-Trihydroxyisoflavone

24388232-1G 24388232-100MG
MOLE24388232-1GEA 119 EUR
MOLE24388232-1G MOLE24388232-100MG
Genistein
Genistein
Formula: C₁₅H₁₀O₅
MW: 270,24 g/mol
Melting Pt: 297…298 °C
MDL Number: MFCD00016952
CAS Number: 446-72-0
EINECS: 207-174-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR