4-Bromo-2-(trifluoromethyl)benzenesulphonyl chloride ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Bromo-2-(trifluoromethyl)benzene-1-sulphonyl chloride

L19515.03 L19515.06
L19515.03EA 57.8 EUR
L19515.03 L19515.06
4-Bromo-2-(trifluoromethyl)benzenesulphonyl chloride ≥97%
4-Bromo-2-(trifluoromethyl)benzenesulphonyl chloride

Purity: 97% CAS No.: 176225-10-8 Molecular Formula: C7H3BrClF3O2S Formula Weight: 323.52 Flash Point: 110°(230°F) Melting Point: 54-58° UN No.: 3261 MDL No.: MFCD03094392

Formula: C₇H₃BrClF₃O₂S
MW: 323,52 g/mol
Melting Pt: 54…58 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD03094392
CAS Number: 176225-10-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR