4-Chloro-3-fluorophenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Chloro-3-fluorobenzeneboronic acid, (4-Chloro-3-fluorophenyl)metaboric acid, (4-Chloro-3-fluorophenyl)boronic acid, Dihydroxy(4-chloro-3-fluorophenyl)borane

H53325.03 H53325.06
H53325.03EA 52.9 EUR
H53325.03 H53325.06
4-Chloro-3-fluorophenylboronic acid ≥97%
4-Chloro-3-fluorophenylboronic acid

Purity: 97% CAS No.: 137504-86-0 Molecular Formula: C6H5BClFO2 Formula Weight: 174.37 Melting Point: 198-204° MDL No.: MFCD01319010

Formula: C₆H₅BClFO₂
MW: 174,37 g/mol
Melting Pt: 198…204 °C
Storage Temperature: Ambient
MDL Number: MFCD01319010
CAS Number: 137504-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR