4-Chloro-3-nitrobenzenesulphonyl chloride ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Chloro-3-nitrobenzenesulphonyl Chloride, 4-Chloro-3-nitrobenzene-1-sulphonyl chloride

B21632.06 B21632.14 B21632.22
B21632.06EA 34.1 EUR
B21632.06 B21632.14 B21632.22
4-Chloro-3-nitrobenzenesulphonyl chloride ≥98%
4-Chloro-3-nitrobenzenesulphonyl chloride

Purity: 98% CAS No.: 97-08-5 Molecular Formula: C6H3Cl2NO4S Formula Weight: 256.07 Melting Point: 58-61° Beilstein Reference: 1978600 UN No.: 3261 MDL No.: MFCD00007440

Formula: C₆H₃Cl₂NO₄S
MW: 256,07 g/mol
Melting Pt: 58…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00007440
CAS Number: 97-08-5
EINECS: 202-558-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR