2-Fluoroisonicotinonitrile ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-Cyano-2-fluoropyridine
H25938.14
H25938.06
H25938.03
H25938.14EA
192
EUR
H25938.14
H25938.06
H25938.03
2-Fluoroisonicotinonitrile ≥97%
2-Fluoroisonicotinonitrile
Purity: 97%
CAS No.: 3939-14-8
Molecular Formula: C6H3FN2
Formula Weight: 122.10
Melting Point: 31-33°
MDL No.: MFCD09027299
Formula:
C₆H₃FN₂ Melting Pt: 31…33 °C Storage Temperature: Ambient |
MDL Number:
MFCD09027299 CAS Number: 3939-14-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...