4-Fluorothiobenzamide ≥97%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Fluorobenzothioamide

H32093.03 H32093.06
H32093.03EA 34.7 EUR
H32093.03 H32093.06
4-Fluorothiobenzamide ≥97%
4-Fluorothiobenzamide

Purity: 97% CAS No.: 22179-72-2 Molecular Formula: C7H6FNS Formula Weight: 155.19 Melting Point: 146-148° UN No.: 2811 MDL No.: MFCD01313285

Formula: C₇H₆FNS
MW: 155.2 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD01313285
CAS Number: 22179-72-2
EINECS: 244-820-9
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR