4',7-Dimethoxyisoflavone ≥97%
Supplier: Thermo Fisher Scientific
Synonyms:
7-Methoxy-3-(4-methoxyphenyl)chromen-4-one, 7-Methoxy-3-(4-methoxyphenyl)-4-benzopyrone, 4',7-Dimethoxyisoflavone
L13838.04
L13838.MC
L13838.04EA
0
EUR
L13838.04
L13838.MC
4',7-Dimethoxyisoflavone ≥97%
4',7-Dimethoxyisoflavone
Purity: 97%
CAS No.: 1157-39-7
Molecular Formula: C17H14O4
Formula Weight: 282.30
Melting Point: 161-163°
Beilstein Reference: 265550
Formula:
C₁₇H₁₄O₄ Melting Pt: 161…163 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075889 CAS Number: 1157-39-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...