4-Methoxythiobenzamide ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
4-Methoxybenzothioamide
L10945.06
L10945.03
L10945.06EA
176
EUR
L10945.06
L10945.03
4-Methoxythiobenzamide ≥98%
4-Methoxythiobenzamide
Purity: 98%
CAS No.: 2362-64-3
Molecular Formula: C8H9NOS
Formula Weight: 167.23
Melting Point: 145-147°
Beilstein Reference: 1072444
Formula:
C₈H₉NOS MW: 167.23 g/mol Boiling Pt: 282 Melting Pt: 145…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040993 CAS Number: 2362-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...